missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (R338), DyLight 594 , Novus Biologicals™
Shop All Bio Techne ProductsDescription
Human Respiratory Syncytial Virus A2 Fusion Protein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | Human Respiratory Syncytial Virus A2 Fusion Protein |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | R338 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HRSVgp08, RSV, RSV fusion protein |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (RSV) Fusion glycoprotein/ RSV-F (AAB59858.1; Met1-Thr529) and was produced using recombinant antibody technology. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?