missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (R338), Alexa Fluor™ 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89761AF350
453.05 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Human Respiratory Syncytial Virus A2 Fusion Protein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| Human Respiratory Syncytial Virus A2 Fusion Protein | |
| Monoclonal | |
| Alexa Fluor 350 | |
| HRSVgp08, RSV, RSV fusion protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (RSV) Fusion glycoprotein/ RSV-F (AAB59858.1; Met1-Thr529) and was produced using recombinant antibody technology. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| R338 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction