missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Renin Quantikine ELISA Kit

Product Code. 18742434
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18742434

Brand: R&D Systems™ DREN00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This assay detects pro- and mature Renin.

The Human Renin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Renin in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul), urine (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 14.8 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human Renin. This assay recognizes both the Pro- and mature versions of Recombinant Human Renin.
For Use With (Application) ELISA
Gene ID (Entrez) 5972
Gene Symbol REN
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, REN, renin, renin precursor, renal
Target Renin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.