missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Renin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£183.00 - £537.00
Specifications
| Antigen | Renin |
|---|---|
| Clone | 411506 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723507
![]() |
R&D Systems
MAB40901-SP |
25 μg |
£183.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761237
|
R&D Systems
MAB40901-100 |
100 μg |
£537.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Renin Monoclonal antibody specifically detects Renin in Human samples. It is validated for ELISASpecifications
| Renin | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, REN, renin, renin precursor, renal | |
| Mouse myeloma cell line NS0-derived recombinant human Renin, Leu24-Arg406, Accession # P00797 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 411506 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00797 | |
| 5972 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title