missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Renalase Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5350-SP
This item is not returnable.
View return policy
Description
Renalase Polyclonal specifically detects Renalase in Human samples. It is validated for Western Blot.
Specifications
| Renalase | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C10orf59, C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase, RNLS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55328 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_001026879 | |
| RNLS | |
| E. coli-derived recombinant human Renalase Ala2-Ile342 Accession # NP_001026879 | |
| 25 μg | |
| Primary | |
| Detects human Renalase in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction