missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Relaxin-3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB31071
This item is not returnable.
View return policy
Description
Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Relaxin-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| H3, INSL7, M3, relaxin 3, Relaxin3, RLN3, RXN3, ZINS4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 117579 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization | |
| 332105 | |
| Western Blot 1 ug/mL, Neutralization 1-5 ug/mL | |
| Q8WXF3 | |
| RLN3 | |
| E. coli-derived recombinant human Relaxin-3 Arg26-Cys142 Accession # Q8WXF3 | |
| 500 μg | |
| Primary | |
| Recognizes the pro and mature forms of human Relaxin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with the pro forms of recombinant human (rh) Relaxin-1 or rhRelaxin-2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction