missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Relaxin-2 Recombinant Protein, PeproTech®
Description
Recombinant Human Relaxin-2 is a non-glycosylated, 6.0 kDa, disulfide-linked, heterodimeric protein consisting of a 24 amino acid A-chain, and a 29 amino acid B-chain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Relaxin 1 (also referred to as Relaxin or RLXH1) is a peptide hormone produced by the corpora lutea of the ovary during pregnancy in many mammalian species, including human. The secretion of the hormone into the blood stream just before parturition results in a marked softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the birth process. By inhibiting uterine contractions, Relaxin 1 may influence the timing of parturition. Like insulin, Relaxin 1 consists of two peptide chains, A and B, covalently linked by disulfide bonds. By further analogy to insulin, the two peptides are synthesized as a single-chain precursor polypeptide with the B chain at the amino-terminus. The gene that encodes the human Relaxin 1 protein maps to chromosome 9. Relaxin 2, a related protein, is selectively expressed in the ovary during pregnancy. The gene that encodes the human Relaxin 2 protein also maps to chromosome 9.
Specifications
Specifications
| Accession Number | P04090 |
| For Use With (Application) | Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 6019 |
| Molecular Weight (g/mol) | 6 kDa |
| Name | Human Relaxin-2 |
| Purification Method | Purified |
| Quantity | 25 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?