missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reg4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£159.00 - £458.00
Specifications
| Antigen | Reg4 |
|---|---|
| Clone | 200214 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732357
![]() |
R&D Systems
MAB1379-SP |
25 μg |
£159.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18162481
|
R&D Systems
MAB1379 |
500 μg |
£458.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reg4 Monoclonal specifically detects Reg4 in Human samples. It is validated for Western Blot.Specifications
| Reg4 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BYZ8 | |
| 83998 | |
| E. coli-derived recombinant human Reg4 Asp23-Pro158 Accession # Q9BYZ8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Reg4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Reg1, rmReg2, recombinant rat (rr) Reg3B, rmReg3A, rrReg3A, or rhReg1B. |
| 200214 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gastrointestinal secretory protein, GISP, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELP, RELPREG-like protein | |
| REG4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title