missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reg1A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4937
This item is not returnable.
View return policy
Description
Reg1A Monoclonal specifically detects Reg1A in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Reg1A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P05451 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human Reg1A in Western blots. Shows 5% cross-reactivity with recombinant human (rh) Reg1B and no cross-reactivity with rhReg3A, recombinant mouse Reg1, or recombinant rat Reg1. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 431202 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| REG1A | |
| E. coli-derived recombinant human Reg1A Gln23-Asn166 Accession # P05451 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction