missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human REEP1 (aa 153-200) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63515 (PA5-63515. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
REEP1 belongs to the DP1 family and may enhance the cell surface expression of odorant receptors.
Specifications
Specifications
| Accession Number | Q9H902 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 65055 |
| Name | Human REEP1 (aa 153-200) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C2orf23; D6Ertd253e; HMN5B; rcG_56072; receptor accessory protein 1; receptor expression-enhancing protein 1; receptor expression-enhancing protein 1-like; Reep1; RGD1305230; si:dkey-21a6.4; Spastic paraplegia 31 protein; SPG31; Yip2a |
| Common Name | REEP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction