missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant VEGF-121, ACF
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Vascular endothelial growth factor (VEGF-121) is a naturally-occurring VEGF-A splice variant involved in embryonic vasculogenesis and angiogenesis
- VEGF binds to VEGF receptor (VEGFR)-1 and VEGFR-2, and activates Raf/MEK/ERK and PI3K/AKT pathways
- VEGF-121 is released as a freely diffusible protein by a variety of normal and transformed cells
- It plays an important role in neurogenesis both in vitro and in vivo (Storkebaum et al.) It has neurotrophic effects on neurons of the central nervous system and promotes growth and survival of dopaminergic neurons and astrocytes
- VEGF also promotes growth and survival of vascular endothelial cells, monocyte chemotaxis, and colony formation by granulocyte-macrophage progenitor cells (Ferrara et al.) VEGF-121 contains two polypeptide chains of 121 amino acids each
- This product is animal component-free
Specifications
Specifications
| Name | MGC70609, MVCD-1, Vascular endothelial growth factor, Vascular endothelial growth factor 2, Vascular endothelial growth factor A, Vascular permeability factor, VEGFA, VPF |
| Quantity | 10 μg |
| Cell Type | Angiogenic Cells; Endothelial Cells; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction