missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant TPO
Description
- Thrombopoietin (TPO) is a key regulator of megakaryocytopoiesis and thrombopoiesis in vitro and in vivo
- TPO stimulates the proliferation and maturation of megakaryocytes and has an important role in regulating the level of circulating platelets in vivo (Bartley et al.; de Sauvage et al.; Foster et al.; Sohma et al.) TPO also promotes the survival, self-renewal, and expansion of hematopoietic stem cells and primitive multilineage progenitor cells
- It is commonly used with other cytokines such as stem cell factor (SCF) and Flt3/Flk-2 ligand to promote expansion of primitive hematopoietic cells in culture (Hitchcock & Kaushansky)
- The TPO receptor, c-Mpl, is expressed at all stages of megakaryopoiesis, from hematopoietic stem and progenitor cells to mature platelets (Ng et al.)
Specifications
Specifications
| Name | c-MPL ligand, Megakaryocyte colony-stimulating factor, MGDF, MKCSF, ML, MPLLG, THCYT1, THPO |
| Quantity | 1000 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥90% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?