missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant TPO

Product Code. 17169841
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
25 μg
Unit Size:
1000µg
100µg
25µg
This item is not returnable. View return policy

Product Code. 17169841

Brand: STEMCELL Technologies 78210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thrombopoietin

  • Thrombopoietin (TPO) is a key regulator of megakaryocytopoiesis and thrombopoiesis in vitro and in vivo
  • TPO stimulates the proliferation and maturation of megakaryocytes and has an important role in regulating the level of circulating platelets in vivo (Bartley et al.; de Sauvage et al.; Foster et al.; Sohma et al.) TPO also promotes the survival, self-renewal, and expansion of hematopoietic stem cells and primitive multilineage progenitor cells
  • It is commonly used with other cytokines such as stem cell factor (SCF) and Flt3/Flk-2 ligand to promote expansion of primitive hematopoietic cells in culture (Hitchcock & Kaushansky)
  • The TPO receptor, c-Mpl, is expressed at all stages of megakaryopoiesis, from hematopoietic stem and progenitor cells to mature platelets (Ng et al.)

Specifications

Name c-MPL ligand, Megakaryocyte colony-stimulating factor, MGDF, MKCSF, ML, MPLLG, THCYT1, THPO
Quantity 25 μg
Cell Type Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
Purity or Quality Grade ≥90%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.