missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant TNF-beta
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Tumor necrosis factor-beta (TNF-β), also known as lymphotoxin-alpha, is a member of the TNF family
- It is a pro-inflammatory cytokine that activates NF-kB, MAPK, and PI3K/AKT pathways upon binding to TNF receptors 1 and 2
- It is produced mainly by T cells, though other cells can express TNF-β at lower levels (Chu)
- TNF-β is involved in autoimmune disorders, lymph node development, and mediating the inflammatory demyelination process (Seleznik et al.; McCarthy et al.)
Specifications
Specifications
| Name | LT-α, Lymphotoxin-alpha, TNF-beta, TNFSF1, Tumor necrosis factor-β |
| Quantity | 10 μg |
| Cell Type | T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction