missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant TNF-alpha

Product Code. p-7217606
Encompass_Preferred
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 17169491

Brand: STEMCELL Technologies 78068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tumor necrosis factor alpha

  • Tumor necrosis factor-α (TNF-α) is a pro-inflammatory cytokine that activates NF-kB, MAPK, and PI3K/AKT pathways
  • Activated T cells and macrophages are the primary producers of TNF-α in response to inflammation and infectious conditions
  • Many other cell types have been shown to produce TNF-α, among them B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts
  • TNF-α has cytotoxic effects on cancer cells in vitro by stimulating anti-tumor immuno- suppressive responses
  • TNF-α stimulates expression of E- and P-selectins, thus facilitating adhesion of neutrophils, monocytes, and memory T cells to activated platelets and endothelial cells (Zelová & Hosek)
  • Other effects of TNF-α include vasodilatation and edema formation

Specifications

Name Cachectin, Cachexin, Cytotoxin, DIF, Necrosin, TNF, TNF-α, TNFSF2, Tumor necrosis factor-α
Quantity 50 μg
Cell Type Airway Cells; B Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Neurons; NK Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.