missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant TGF-beta 3, ACF

Product Code. 17149711
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
4 x 250 μg
Unit Size:
100µg
10µg
Pack of 4
This item is not returnable. View return policy

Product Code. 17149711

Brand: STEMCELL Technologies 78156

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Transforming growth factor beta 3, animal component-free

  • Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes
  • TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.) TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.) TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.) This product is animal component-free

Specifications

Name ARVD, FLJ16571, TGFB3, TGF-b3, Transforming growth factor beta-3
Quantity 10 μg
Cell Type Mesenchymal Cells, PSC-Derived; Mesenchymal Stem and Progenitor Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.