missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant Shh (C24II)
Description
- Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho & Scott)
- It is expressed during early embryogenesis in a variety of tissues (Finco et al.) During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocytes precursors and neural crest cells and stimulates proliferation, differentiation and axon growth (Ho & Scott; Martí)
- Shh also regulates T cell differentiation and activation (Crompton et al.) Recombinant human Shh (C24II; rhShh) is a fully biologically active molecule
Specifications
Specifications
| Name | HHG-1, HLP3, HPE3, MCOPCB5, SMMCI, Sonic hedgehog, TPT, TPTPS |
| Quantity | 50 μg |
| Cell Type | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?