missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant SCF, ACF

Product Code. p-7217683
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17119711

Brand: STEMCELL Technologies 78155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stem cell factor, animal component-free

  • Stem cell factor (SCF) is an early-acting cytokine that plays a pivotal role in the regulation of embryonic and adult hematopoiesis
  • SCF promotes cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy
  • Together with other cytokines such as thrombopoietin and Flt3/Flk-2 Ligand, SCF is commonly used to promote expansion of primitive hematopoietic stem cells and multipotent progenitor cells in culture
  • In synergy with various growth factors, including IL-2, IL-3, IL-6, IL-7, G-CSF, and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells
  • SCF is also a primary growth and activation factor for mast cells and eosinophils
  • This product is animal component-free

Specifications

Name Kit ligand, Mast cell growth factor, Steel factor, Stem cell factor
Quantity 10 μg
Cell Type Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.