Learn More
STEMCELL Technologies Human Recombinant RANTES (CCL5)
Product Code. 17181046
Encompass_Preferred
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Product Code. 17181046
Brand: STEMCELL Technologies 78099
Regulated upon activation, normal T cell expressed and secreted
  • RANTES (regulated upon activation, normal T cell expressed and secreted), also known as CCL5, is a member of the CC family of chemokines and is able to recruit leukocytes to sites of inflammation
  • RANTES is secreted by T lymphocytes, macrophages, platelets, synovial fibroblasts, tubular epithelium, and certain types of tumor cells
  • This chemokine exerts its effect by interacting with the chemokine receptors CCR1, CCR3, CCR4, and CCR5
  • RANTES plays an active role in recruiting a variety of leukocytes into inflammatory sites, including T cells, macrophages, eosinophils, and basophils
  • In collaboration with certain cytokines that are released by T cells such as IL-2 and IFN-γ, RANTES also induces the activation and proliferation of NK cells to generate CC chemokine-activated killer cells, which are highly cytolytic
Specifications
Name D17S136, Eosinophil chemotactic cytokine, Regulated upon activation normally T-expressed and secreted, SIS-delta, Small-inducible cytokine A5, T-cell specific protein p288
Quantity 20 μg
Cell Type B Cells; Dendritic Cells; Mesenchymal Stem and Progenitor Cells; Monocytes; NK Cells; Other; T Cells; T Cells, CD4+; T Cells, CD8+
Purity or Quality Grade ≥95%