missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant Prolactin

Product Code. 17169561
Encompass_Preferred
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 17169561

Brand: STEMCELL Technologies 78098

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prolactin

  • Prolactin is a peptide hormone of pituitary origin and is well known for the stimulation, initiation, and maintenance of lactation
  • Prolactin is also known to regulate mammary gland development and immune system, and has an essential role in metabolism and behavioral modification (Bernichtein et al.) Prolactin actions are mediated by the prolactin receptor (PRLR), which is a single-chain transmembrane protein composed of an extracellular, transmembrane, and intracellular domain
  • It has also been indicated that high to normal circulating levels of prolactin increase breast cancer risk, and dopamine agonists have been suggested to be effective for treatment (Bernichtein et al.) Prolactin levels have been reported to be higher in multiple sclerosis patients, which promotes B cell autoreactivity (Correale et al.) It has been shown that administration of recombinant prolactin after bone marrow transplantation in mice promotes immune and myeloid reconstitution (Sun et al.)

Specifications

Name Luteotropin, Luteotropic hormone, Mammotropin, PRL
Quantity 10 μg
Cell Type Adipocytes; Lymphocytes; Mammary Cells; Neural Stem and Progenitor Cells; Neurons
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.