missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant PDGF-AB
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (Fretto et al.; Westermark & Heldin)
- PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta
- It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.) PDGF is a potent mitogen for cells of mesenchymal origin- like fibroblasts, glial cells, and vascular smooth muscle cells
- PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.) It has been shown that PDGF-AB together with 5-Azacytidine (Catalog #72012), induces the conversion of mature bone and fat cells into tissue-regenerative multipotent stem cells (Chandrakanthan et al.)
Specifications
Specifications
| Name | GDGF, Glioma-derived growth factor, ODGF, Osteosarcoma-derived growth factor, Platelet-derived growth factor-AB |
| Quantity | 100 μg |
| Cell Type | Mesenchymal Cells, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Osteoblasts; Other |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction