missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant PDGF-AA

Product Code. 17169551
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
This item is not returnable. View return policy

Product Code. 17169551

Brand: STEMCELL Technologies 78095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platelet-derived growth factor AA

  • Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB)
  • PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta
  • It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT
  • PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells
  • PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues
  • It has been suggested that PDGF-AA is an important autocrine regulator of vascular endothelial growth factor (VEGF) expression in non-small cell lung carcinomas

Specifications

Name GDGF, Glioma-derived growth factor, ODGF, Osteosarcoma-derived growth factor, Platelet-derived growth factor-AA
Quantity 10 μg
Cell Type Mesenchymal Cells, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Other
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.