missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant Oncostatin M
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- Oncostatin M (OSM) is a member of interleukin 6 (IL-6) family of cytokines and bears close resemblance to leukemia-inhibitory factor (LIF) and granulocyte colony-stimulating factor (G-CSF) in amino acid sequence and its modulation of differentiation in a variety of cell types
- OSM signals through type I receptor (consisting of gp130 and LIF receptor (LIFR)) and type II receptor (consisting of gp130 and OSM receptor (OSMR)), which eventually activate the JAK/STAT pathway
- OSM is primarily produced by activated T cells and monocytes, and also by activated macrophages, neutrophils, mast cells, and dendritic cells
- OSM is also produced within the bone microenvironment by cells of both hematopoietic and mesenchymal origin including osteocytes and osteoblasts
Specifications
Specifications
| Name | OSM |
| Quantity | 50 μg |
| Cell Type | Airway Cells; B Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Lymphocytes; Neurons; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction