missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant NGF-beta

Product Code. 17109551
Encompass_Preferred
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
This item is not returnable. View return policy

Product Code. 17109551

Brand: STEMCELL Technologies 78092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nerve growth factor beta

  • Nerve growth factor (NGF)-beta is a prototypical member of the neurotrophin family and has a role in the survival and growth of neural cells, regulating cell growth, promoting differentiation into neurons, and neuron migration
  • The beta subtype of NGF is biologically active in comparison to the alpha-2 and gamma-2 subtypes
  • NGF-beta in its secreted form can bind to tyrosine kinase A (TrkA) receptor with high affinity and to p75 (NTR) with low affinity (Levi & Alemà; Sofroniew et al.) NGF has been shown to possess pro-inflammatory and pro-fibrogenic properties (Micera et al.) It has also been shown that overexpression of NGF-beta promotes differentiation of bone marrow mesenchymal stem cells into neurons through regulation of AKT and MAPK pathways (Yuan et al.)

Specifications

Name Beta-nerve growth factor, beta-NGF, nerve growth factor (beta polypeptide)
Quantity 20 μg
Cell Type Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.