missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant Myostatin

Product Code. 17129611
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17129611 10 μg 10µg
17139611 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17129611 Supplier STEMCELL Technologies Supplier No. 78119

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Myostatin

  • Myostatin is a member of transforming growth factor beta (TGF-β) superfamily that signals by binding to activin type II receptor, resulting in the recruitment of either ALK3 or ALK4 coreceptor, and activation of SMAD2/3 (Kondás et al.) It is released by myocytes and inhibits muscle growth and ability to regenerate (Lee & Lee)
  • Myostatin appears to affect the lipid catabolic metabolism of adipocytes and inhibits preadipocyte differentiation, as demonstrated in the 3T3-L1 cell line (Li et al.)

Specifications

Name GDF-8, Growth differentiation factor 8, MSTN78
Quantity 10 μg
Cell Type Mesenchymal Stem and Progenitor Cells
Purity or Quality Grade ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.