missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IP-10 (CXCL10)
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Interferon gamma-inducible protein (IP) 10 or CXCL10 is a member of the CXC chemokine family
- It binds CXCR3 activating ERK1/2, p38/MAPK, JNK, and PI3-kinase/AKT signaling pathways, inducing intracellular calcium influx, DNA synthesis, cell proliferation, and chemotaxis
- IP-10 regulates innate and adaptive immune responses by affecting the function of activated T cells, natural killer cells, inflammatory dendritic cells, macrophages, and B cells
- IP-10 is produced by leukocytes, activated neutrophils, eosinophils, monocytes, epithelial cells, endothelial cells, fibroblasts, and keratinocytes in response to IFN-gamma
- IP-10 has been implicated in a wide range of diseases, including infectious diseases and chronic inflammatory and autoimmune diseases, as well as in tumor formation (Liu et al.)
Specifications
Specifications
| Name | 10 kDa interferon-gamma-induced protein, C7, crg-2, Chemokine (C-X-C motif) ligand 10, CXCL10, Gamma-IP-10, gIP-10, IFI10, INP10, mob1, SCYB10, Small inducible cytokine B10 |
| Quantity | 50 μg |
| Cell Type | Monocytes; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction