missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-3, ACF
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Interleukin 3 (IL-3) is a species-specific pleiotropic cytokine that promotes the survival and proliferation of pluripotent hematopoietic stem cells and lineage-committed progenitor cells and their differentiation into mature cells of most lineages, including basophils, neutrophils, eosinophils, macrophages, dendritic cells, erythrocytes, and megakaryocytes
- IL-3 is produced by activated T cells and has a physiological role in inflammation and allergies by promoting the secretion of inflammatory mediators such as histamine, IL-4, and IL-6 by basophils and eosinophils
- The IL-3 receptor consists of a unique alpha subunit (CD123) and a beta common subunit (βc or CD131) that is shared with the receptors for IL-5 and GM-CSF, and is the principal signal transduction subunit for these cytokines
- This product is animal component-free
Specifications
Specifications
| Name | HCGF, Interleukin-3, MCGF, Multi-CSF, P-cell stimulation factor |
| Quantity | 10 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction