missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant IL-2 (CHO-expressed)

Product Code. 15881468
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
50 μg
Unit Size:
1000µg
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15881468

Brand: STEMCELL Technologies 78036.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 2

  • Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor
  • It binds to heterotrimeric receptors consisting of CD25, CD122, and CD132
  • Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival (Ma et al.) The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts
  • IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ regulatory T cells, and affects development of follicular helper T cells
  • IL-2 also controls inflammation by inhibiting Th17 differentiation
  • Human Recombinant IL-2 has a serine substitute for cysteine at position 126

Specifications

Name Aldesleukin, Interleukin-2, T cell growth factor, TCGF
Quantity 10 μg
Cell Type Hematopoietic Stem and Progenitor Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.