missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant IL-12

Product Code. 17109391
Encompass_Preferred
Click to view available options
Quantity:
1000 μg
25 μg
5 μg
Unit Size:
1000µg
25µg
5µg
This item is not returnable. View return policy

Product Code. 17109391

Brand: STEMCELL Technologies 78027.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 12

  • Interleukin 12 (IL-12p70) is a heterodimeric cytokine composed of p35 and p40 subunits
  • IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils, and B cells in response to bacterial products and cytokines such as IFN-γ
  • The IL-12 receptor is expressed on T, NK, and dendritic cells
  • Upon binding, IL-12 initiates signaling via the JAK/STAT signaling pathway and stimulates NK, B, and T cells to produce IFN-γ (Watford et al.) It also regulates cytokine synthesis, proliferation of T and NK cells, and stimulates differentiation of CD4+ and CD8+ T cells (Germann & Rüde)
  • Mice that are deficient in IL-12 are susceptible to many intracellular pathogens and have impaired IFN-γ secretion, Th1 differentiation, and NK cytolytic activity; however, Th2 development and IL-4 production are enhanced (Watford et al.)

Specifications

Quantity 1000 μg
Cell Type B Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells; T Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.