missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-12
Click to view available options
Quantity:
1000 μg
25 μg
5 μg
Unit Size:
1000µg
25µg
5µg
Description
- Interleukin 12 (IL-12p70) is a heterodimeric cytokine composed of p35 and p40 subunits
- IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils, and B cells in response to bacterial products and cytokines such as IFN-γ
- The IL-12 receptor is expressed on T, NK, and dendritic cells
- Upon binding, IL-12 initiates signaling via the JAK/STAT signaling pathway and stimulates NK, B, and T cells to produce IFN-γ (Watford et al.) It also regulates cytokine synthesis, proliferation of T and NK cells, and stimulates differentiation of CD4+ and CD8+ T cells (Germann & Rüde)
- Mice that are deficient in IL-12 are susceptible to many intracellular pathogens and have impaired IFN-γ secretion, Th1 differentiation, and NK cytolytic activity; however, Th2 development and IL-4 production are enhanced (Watford et al.)
Specifications
Specifications
| Name | CTL maturation factor, Cytotoxic lymphocyte maturation factor, IL-12p70, Interleukin-12, Natural killer cell stimulatory factor, NKSF |
| Quantity | 25 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction