missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant IL-11

Product Code. p-7217586
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17119381

Brand: STEMCELL Technologies 78025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin 11

  • Interleukin 11 (IL-11) is a pleiotropic cytokine with effects on various tissues including the bone marrow, brain, and intestinal mucosa
  • It belongs to the IL-6 family of cytokines that share a common signal transducer, gp130
  • IL-11 induces the proliferation of hematopoietic stem cells and megakaryocytic progenitor cells, the maturation of megakaryocytes, and the production of platelets
  • IL-11 is produced by a variety of cell types including hematopoietic cells, mesenchymal cells, epithelial cells, and neuronal cells
  • It was first cloned from a cDNA library of the human bone marrow-derived stromal cell line KM-102
  • The binding of IL-11 to its receptor induces heterodimerization with the gp130 subunit and activation of JAK tyrosine kinases
  • IL-11 was the first pharmacologic agent approved for the treatment of chemotherapy-induced thrombocytopenia

Specifications

Name Adipogenic inhibitory factor, AGIF, Interleukin-11, Oprelvekin
Quantity 100 μg
Cell Type B Cells; Hematopoietic Stem and Progenitor Cells; Other; Pluripotent Stem Cells; T Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.