missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-11
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
- Interleukin 11 (IL-11) is a pleiotropic cytokine with effects on various tissues including the bone marrow, brain, and intestinal mucosa
- It belongs to the IL-6 family of cytokines that share a common signal transducer, gp130
- IL-11 induces the proliferation of hematopoietic stem cells and megakaryocytic progenitor cells, the maturation of megakaryocytes, and the production of platelets
- IL-11 is produced by a variety of cell types including hematopoietic cells, mesenchymal cells, epithelial cells, and neuronal cells
- It was first cloned from a cDNA library of the human bone marrow-derived stromal cell line KM-102
- The binding of IL-11 to its receptor induces heterodimerization with the gp130 subunit and activation of JAK tyrosine kinases
- IL-11 was the first pharmacologic agent approved for the treatment of chemotherapy-induced thrombocytopenia
Specifications
Specifications
| Name | Adipogenic inhibitory factor, AGIF, Interleukin-11, Oprelvekin |
| Quantity | 100 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Other; Pluripotent Stem Cells; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction