missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-1 beta
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
- Interleukin 1 beta (IL-1β) is synthesized as an inactive precursor protein or pro-IL-1β
- This precursor is cleaved intracellularly by caspase 1 (IL-1β convertase) to form the active form of the protein that is later secreted (Allan et al.) IL-1β binds to IL-1 receptor and activates intracellular signaling via the MAPK or NF-κB pathway
- IL-1β is released by monocytes, tissue macrophages, and dendritic cells in response to infection or injury and induces expression of acute-phase proteins
- It also promotes the infiltration of inflammatory and immunocompetent cells from the circulation into the extravascular space and affected tissues, by stimulating the expression of adhesion molecules on endothelial cells
- IL-1β also affects other immune cells; for example, it co-stimulates T cell functions together with an antigen or mitogen
- It also stimulates Th17 differentiation and B cell proliferation in an IL-6-dependent manner
Specifications
Specifications
| Name | Catabolin, Endogenous pyrogen, EP, Interleukin-1 beta, LAF, LEM, Leukocyte endogenous mediator, Lymphocyte-activating factor, MCF, Mononuclear cell factor |
| Quantity | 100 μg |
| Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; T Cells; T Cells, CD4+; T Cells, CD8+ |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction