missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant Heregulin-beta 1
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- Heregulin-beta 1 also known as neuregulin-1 (NRG-1) is a member of the epidermal growth factor (EGF) family of growth factors and acts as a ligand for ErbB family receptor tyrosine kinases
- Heregulin/neuregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3, and NRG4)
- Heregulin-beta 1 plays an important role during the development of the nervous system, heart, and mammary glands
- Heregulin-beta 1 is expressed in neuronal cells, and modulates cell growth and differentiation of the cells during development and wound healing
- It has been implicated through in vivo and in vitro studies that heregulin-beta 1/ErbB signaling is crucial for multiple aspects of cardiovascular development and protects the heart from ischemic injury
- Heregulin-beta 1 also promotes invasiveness and metastasis of breast cancer cells
Specifications
Specifications
| Quantity | 1000 μg |
| Cell Type | Cancer Cells and Cell Lines; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction