missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant Heregulin-beta 1

Product Code. 17129501
Encompass_Preferred
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 17129501

Brand: STEMCELL Technologies 78071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Heregulin-beta 1

  • Heregulin-beta 1 also known as neuregulin-1 (NRG-1) is a member of the epidermal growth factor (EGF) family of growth factors and acts as a ligand for ErbB family receptor tyrosine kinases
  • Heregulin/neuregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3, and NRG4)
  • Heregulin-beta 1 plays an important role during the development of the nervous system, heart, and mammary glands
  • Heregulin-beta 1 is expressed in neuronal cells, and modulates cell growth and differentiation of the cells during development and wound healing
  • It has been implicated through in vivo and in vitro studies that heregulin-beta 1/ErbB signaling is crucial for multiple aspects of cardiovascular development and protects the heart from ischemic injury
  • Heregulin-beta 1 also promotes invasiveness and metastasis of breast cancer cells

Specifications

Name Acetylcholine receptor-inducing activity, ARIA, Breast cancer cell differentiation factor p45, Glial growth factor, Heregulin Neu differentiation factor, HRG, HRG1, HRG1-beta 1, Neuregulin-1, NRG1, NRG1-beta 1, Sensory and motor neuron-derived factor
Quantity 10 μg
Cell Type Cancer Cells and Cell Lines; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.