missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant HBEGF
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is a member of the EGF family (Nishi & Klagsbrun)
- HBEGF promotes blastocyst adhesion to the uterine wall (Iwamoto & Mekada)
- It also plays a role in smooth muscle cell hyperplasia and brain injury (Nishi & Klagsburn)
- HBEGF produced by CD4+ T cells promotes wound healing by stimulating migration and proliferation of keratinocytes, fibroblasts, and smooth muscle cells (Blotnick et al.) It binds to EGFR, ErbB4, ErbB2, and ErbB3, activating the PI3K/AKT signaling cascade (Iwamoto & Mekada)
- HBEGF is produced in a variety of cells, where it contributes to physiological and pathological processes
- HBEGF is overexpressed in ovarian, breast, gastric, colorectal, pancreatic, and endometrial cancers, which likely contributes to pathogenesis (Miyata et al.)
Specifications
Specifications
| Name | Diphtheria toxin receptor, DTR, HBEGF, Heparin binding EGF-like growth factor |
| Quantity | 10 μg |
| Cell Type | Fibroblasts; Keratinocytes; Smooth Muscle; T Cells, CD4+ |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction