missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant GDF-5
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
Description
- Growth differentiation factor 5 (GDF-5) is a member of the bone morphogenetic protein (BMP) subclass of the transforming growth factor beta (TGF-β) superfamily
- It binds a receptor complex comprising BMPR1B and BMPR2, which then activates the SMAD signaling pathway (Carreira et al.; Nishitoh et al.; Osório et al.) GDF-5 enhances chondrogenic differentiation of mesenchymal stem cells, skeletogenesis, and dendrite growth during development (Coleman et al.; Francis-West et al.) Studies in rat demonstrated that GDF-5 regulates patterning, neurogenesis, astrogliogenesis, and neuronal specification (Gajavelli et al.; Krieglstein et al.; O'Keeffe et al.)
Specifications
Specifications
| Name | BMP-14, Bone morphogenic protein 14, CDMP-1, Growth differentiation factor 5 |
| Quantity | 50 μg |
| Cell Type | Endoderm, PSC-Derived; Neural Stem and Progenitor Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction