missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant GDF-11
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Description
- Growth differentiation factor 11 (GDF-11) is a member of transforming growth factor beta (TGF-β) family
- It binds to the TGF-β receptors ALK4, ALK5, and ALK7 and activates the SMAD signaling pathway (Ho et al.) GDF-11 regulates the development of the olfactory system, retina, and pancreas, as well as anterior/posterior patterning of the axial skeleton (Lee & Lee)
- GDF-11 is an endocrine factor expressed in skeletal muscle, brain, and dental pulp (Kondás et al.) Studies in mice showed that GDF-11 regulates muscle and cardiac aging, and stimulates neurogenesis by remodeling blood vessels (Katsimpardi et al.; Loffredo et al.; Sinha et al.)
Specifications
Specifications
| Name | BMP-11, Bone morphogenetic protein 11, Growth differentiation factor 11 |
| Quantity | 20 μg |
| Cell Type | Neural Cells, PSC-Derived |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction