missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant G-CSF
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
- Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function
- It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells
- G-CSF causes a range of effects including a transient reduction of SDF-1 expression, the activation of metalloproteases that cleave VCAM-1, and the release of norepinephrine from the sympathetic nervous system, leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery
Specifications
Specifications
| Name | Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin |
| Quantity | 500 μg |
| Cell Type | Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neurons |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction