missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant Fractalkine (CX3CL1)
Click to view available options
Quantity:
1000 μg
25 μg
5 μg
Unit Size:
1000µg
25µg
5µg
Description
- Fractalkine (CX3CL1) is a unique chemokine belonging to the CX3C family, and is characterized by a C-X3-C cysteine motif within the chemokine domain, near the amino terminus of the protein
- The chemokine domain is connected to an extended mucin-like stalk, followed by a transmembrane region, and a C-terminal intracellular domain
- The protein signals through interaction with a single receptor, CX3CR1, expressed on monocytes, natural killer cells, T cells, microglia, and smooth muscle cells
- Fractalkine is upregulated in endothelial cells by inflammatory signals and is synthesized as a membrane-bound molecule that mediates cell migration and adhesion
- Cleavage at the base of the stalk by metalloproteinases generates a soluble chemokine, which functions as a potent chemoattractant of target cells
Specifications
Specifications
| Name | C3Xkine, Chemokine (C-X3-C motif) ligand 1, FKN, Neurotactin, NTN, NTT, SCYD1, Small inducible cytokine subfamily D member 1 |
| Quantity | 25 μg |
| Cell Type | Dendritic Cells; Endothelial Cells; Macrophages; Monocytes; NK Cells; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction