missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-acidic
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Fibroblast growth factor acidic (FGF-acidic), also known as FGF-1, is a potent activator of DNA synthesis, cell proliferation, and chemotaxis and is known to play numerous roles in development, regeneration, and angiogenesis
- FGF-acidic is produced by multiple cell types and is capable of activating all cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin
- It is found in large quantities in the brain, but is also expressed in hepatocytes, vascular smooth muscle cells, neurons of the CNS, skeletal muscle cells, fibroblasts, keratinocytes, endothelial cells, intestinal columnar epithelial cells, and pituitary basophils and acidophils
- FGF-acidic is secreted as a disulfide-linked homodimer and is stored in complex with heparan sulfate, a requirement for its interaction with FGF receptors
Specifications
Specifications
| Name | Acidic fibroblast growth factor, aFGF, ECGF, Endothelial cell growth factor, FGF-1, FGF-alpha, Fibroblast growth factor 1, GLIO703, HBGF-1, Heparin-binding growth factor 1 |
| Quantity | 10 μg |
| Cell Type | Endoderm, PSC-Derived; Endothelial Cells; Epithelial Cells; Fibroblasts; Granulocytes and Subsets; Keratinocytes; Mesoderm, PSC-Derived; Neurons; Smooth Muscle |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction