missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-6
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
Description
- Fibroblast growth factor 6 (FGF-6) is a heparin-binding member of the FGF family, regulators of cell proliferation, differentiation, and function
- FGF-6 binds and signals through the FGF receptors 1c, 2c, and 4 (Ornitz et al.) FGF-6 is a potent mitogen for fibroblasts, vascular endothelial cells, and prostate carcinoma cells (Asada et al.; Pizette et al.; Ropiquet et al.) FGF-6 is primarily expressed in epithelial and mesenchymal cell lineages
- During development, FGF-6 is expressed in skeletal muscle, consistent with its role in muscle differentiation and regeneration (Floss et al.) FGF-6 has also been shown to promote chondrogenesis in embryonic somites in conjunction with transforming growth factor beta 2 (TGF-β2; Grass et al.)
Specifications
Specifications
| Name | Fibroblast growth factor 6, HBGF-6, Heparin-binding growth factor 6, Heparin secretory-transforming protein 2, HST-2, HSTF-2 |
| Quantity | 500 μg |
| Cell Type | Endothelial Cells; Fibroblasts; Mesenchymal Stem and Progenitor Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction