missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-18
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Fibroblast growth factor 18 (FGF-18) is a growth factor and member of the FGF subfamily
- FGF-18 is most similar to FGF-8 and FGF-17, and is secreted in adult lung and developing tissues (Ohbayashi et al.) FGF-18 signals through FGF receptor 3 (FGFR3) to regulate proliferation, differentiation, and matrix production of articular and growth plate chondrocytes in vivo and in vitro (Davidson et al.) FGF-18 has skeletal functions and protects articular cartilage by gene expression profiling and regulates proliferation and differentiation of midline cerebellar structures (Mori et al.) Also, recombinant human FGF-18 has been shown to effectively regulate hair growth (Song et al.)
Specifications
Specifications
| Name | FGFI, Fibroblast growth factor-18, zFGF5 |
| Quantity | 10 μg |
| Cell Type | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction