Learn More
STEMCELL Technologies Human Recombinant FGF-18
Product Code. p-7217772
Encompass_Preferred
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Product Code. 17139421
Brand: STEMCELL Technologies 78041
Fibroblast growth factor 18
  • Fibroblast growth factor 18 (FGF-18) is a growth factor and member of the FGF subfamily
  • FGF-18 is most similar to FGF-8 and FGF-17, and is secreted in adult lung and developing tissues (Ohbayashi et al.) FGF-18 signals through FGF receptor 3 (FGFR3) to regulate proliferation, differentiation, and matrix production of articular and growth plate chondrocytes in vivo and in vitro (Davidson et al.) FGF-18 has skeletal functions and protects articular cartilage by gene expression profiling and regulates proliferation and differentiation of midline cerebellar structures (Mori et al.) Also, recombinant human FGF-18 has been shown to effectively regulate hair growth (Song et al.)
Specifications
Name FGFI, Fibroblast growth factor-18, zFGF5
Quantity 10 μg
Cell Type Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells
Purity or Quality Grade ≥95%