missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-16
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Fibroblast growth factor 16 (FGF-16) is a heparin-binding member of the FGF family
- FGFs possess broad mitogenic and cell survival activities and are expressed during embryonic development
- FGFs act primarily on cells of mesodermal and neuroectodermal origin to regulate diverse physiological functions including angiogenesis, cell growth, pattern formation, embryonic development, metabolic regulation, cell migration, and tissue repair (Goldfarb; Green et al.) In vitro, FGF-16 has been shown to promote the proliferation of brown adipocytes and in rat embryos it is predominantly expressed in these cells
- FGF-16 has also been shown to play a critical role in embryonic heart development and is thought to play a cardioprotective role after birth (Hotta et al.; Lu et al.; Wang et al.)
Specifications
Specifications
| Name | Fibroblast growth factor 16, Metacarpal 4-5 fusion, MF4 |
| Quantity | 10 μg |
| Cell Type | Adipocytes |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction