missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-10 (KGF-2), ACF
Click to view available options
Quantity:
100 μg
1000 μg
25 μg
Unit Size:
1000µg
100µg
25µg
Description
- Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family which is predominantly expressed by mesenchymal fibroblasts during embryonic development
- It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb
- FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix
- FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury and endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells
- This product is animal component-free
Specifications
Specifications
| Name | Fibroblast growth factor 10, Keratinocyte growth factor 2 |
| Quantity | 25 μg |
| Cell Type | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction