missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-10 (KGF-2)
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
500 μg
Unit Size:
1000µg
10µg
500µg
50µg
Description
- Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family predominantly expressed by mesenchymal fibroblasts during embryonic development (Emoto et al.; Igarashi et al.) It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb (Beer et al.) FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix (Emoto et al.; Igarashi et al.) FGF-10 has been shown to mediate epithelial-mesenchymal interactions, which are essential to lung development (Sekine et al; Ware & Matthay)
- FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury (Tong et al.; Ware & Matthay), as well as endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells (Takeuchi et al.)
Specifications
Specifications
| Name | FGFA, Fibroblast growth factor-10, Keratinocyte growth factor-2 |
| Quantity | 500 μg |
| Cell Type | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction