missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant EGFR
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Description
- Epidermal growth factor receptor (EGFR) is a type I transmembrane protein and receptor tyrosine kinase
- EGFR has been shown to bind to some members of the EGF family ligands including EGF, amphiregulin, TGF-α, betacellulin, epiregulin, heparin-binding EGF, and neuregulin-2α
- EGFR ligand binding induces homodimerization, as well as heterodimerization of EGFR with ErbB2 or with ligand-activated ErbB3 or ErbB4 (Schlessinger)
- Dimerization results in kinase activation, phosphorylation, and cell signaling, mediated primarily through MEK/ERF and AKT pathways (Navlonic et al.) EGFR signaling has been shown to regulate cell proliferation, differentiation, motility, and apoptosis
- Elevated levels of EGFR have been correlated with carcinogenesis (Maihle et al.) Protein contains a His-residue tag at the carboxyl end of the polypeptide chain
Specifications
Specifications
| Name | Avian erythroblastic leukemia viral oncogene homolog, Epidermal growth factor receptor, Erb-b2 receptor tyrosine kinase 1, ERBB1, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1, v-erb-b |
| Quantity | 10 μg |
| Cell Type | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction