missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant EGF, ACF
Description
- Epidermal growth factor (EGF) is characterized by high-affinity binding to various EGF receptors (EGFRs) and the production of mitogenic responses (Carpenter & Cohen)
- EGF promotes EGFR dimerization, resulting in activation of downstream pathways including PI3K, ERK1/2, JAK/STAT, β-catenin, and calcium signaling
- EGF is secreted by the gut-associated salivary and Brunner's glands, is found in a variety of body fluids, and stimulates cell proliferation and differentiation in rodent and neonatal human intestine (Wright et al.) Central nervous system stem cells also proliferate in response to the EGF stimulus (Reynolds & Weiss)
- This product is animal component-free
Specifications
Specifications
| Name | Beta-urogastrone, HOMG4, Epidermal growth factor, Pro-epidermal growth factor, URG, Urogastrone |
| Quantity | 100 μg |
| Cell Type | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?