missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant BMPR-1A Fc
Click to view available options
Quantity:
100 μg
1000 μg
20 μg
500 μg
Unit Size:
1000µg
100µg
20µg
500µg
Description
- Bone morphogenetic protein receptor type 1A (BMPR-1A) is a type I receptor of the BMP receptor family of transmembrane serine/threonine kinases (Abe; Fischerauer et al.; it is also known as ALK3
- BMPR-1A is a receptor for BMP-2, BMP-4, GDF-5, and GDF-6
- Binding of BMPs to receptor complexes results in phosphorylation of intracellular SMADs, which go on to regulate transcription (Pan et al.) BMP regulation plays a role in proliferation, differentiation, migration, and apoptosis of endothelial and vascular smooth muscle cells (Abe)
- BMPR-1A deficiency is lethal during gastrulation (Mishina et al.) BMPR-1A is important in chondrogenesis and osteogenesis, along with BMPR-1B (Fischerauer et al.; Jing et al.) This protein contains a C-terminus linker (IEGRMD) to an Fc tag (Human IgG1)
Specifications
Specifications
| Name | Activin receptor-like kinase 3, ACVRLK3, ALK3, BMP type-1A receptor, BRK‐1, CD292, Serine/threonine-protein kinase receptor R5, SKR5 |
| Quantity | 1000 μg |
| Cell Type | Chondrocytes; Endoderm, PSC-Derived; Endothelial Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Osteoblasts; Pluripotent Stem Cells; Smooth Muscle |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction